4,4'-Dibromobiphenyl (PBB No. 15) 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: PBB No. 15

A14139-18 A14139-30
AAA14139-18EA 43.51 USD
AAA14139-18 AAA14139-30
4,4'-Dibromobiphenyl (PBB No. 15) 98+%
4,4'-Dibromobiphenyl (PBB No. 15)

Purity: 98+% CAS No.: 92-86-4 Molecular Formula: C12H8Br2 Formula Weight: 312.02 Melting Point: 164-169° Boiling Point: 355-360° UN No.: 3152 MDL No.: MFCD00000101

Formula: C₁₂H₈Br₂
MW: 312 g/mol
Boiling Pt: 355…360 °C
Melting Pt: 164…169 °C
Density: 1.867 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000101
CAS Number: 92-86-4
EINECS: 202-198-6
UN: 3152
ADR: 9,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR