Acetylacetone 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
ACAC, 2,4-Pentanedione
A14117-0F
A14117-AE
A14117-AP
AAA14117-0FEA
286.3
USD
AAA14117-0F
AAAA14117-AE
AAA14117-AP
Acetylacetone 99%
Acetylacetone
Purity: 99%
CAS No.: 123-54-6
Molecular Formula: C5H8O2
Formula Weight: 100.12
Flash Point: 34°(93°F)
Physical Form: Clear liquid
Density: 0.973
Refractive Index: 1.4520
Appearance: Colorless
Melting Point: -23°
Boiling Point: 140-141°
Beilstein Reference: 741937
Merck Reference: 14,81
UN No.: 2310
MDL No.: MFCD00008787
Formula:
CH₃COCH₂COCH₃ MW: 100.12 g/mol Boiling Pt: 140.4 Melting Pt: -23 Density: 0.973 Flash Pt: 34 °C (93 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008787 CAS Number: 123-54-6 EINECS: 204-634-0 UN: 2310 ADR: 3 III Merck Index: 14,81 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...