2-Nitrobenzamide 98%

Supplier: Thermo Scientific Chemicals
Warning

A14084-09 A14084-18
AAA14084-09EA 30 USD
AAA14084-09 AAA14084-18
2-Nitrobenzamide 98%
2-Nitrobenzamide

Purity: 98% CAS No.: 610-15-1 Molecular Formula: C7H6N2O3 Formula Weight: 166.14 Melting Point: 174-178° Boiling Point: 317° MDL No.: MFCD00007976

Formula: C₇H₆N₂O₃
MW: 166.14 g/mol
Boiling Pt: 317 °C
Melting Pt: 174…178 °C
Storage Temperature: Ambient
MDL Number: MFCD00007976
CAS Number: 610-15-1
EINECS: 210-208-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR