2-Vinylpyridine 97% stabilized

Supplier: Thermo Scientific Chemicals
Danger


Stabilization: Stabilised with 4-tert-Butylpyrocatechol (TBC) ≤ 0.1 %

A14056-AP A14056-AE
AAA14056-APEA 150 USD
AAA14056-AP AAA14056-AE
2-Vinylpyridine 97% stabilized
2-Vinylpyridine

Purity: 97% CAS No.: 100-69-6 Molecular Formula: C7H7N Formula Weight: 105.14 Storage Temperature: 2°C to 8°C Flash Point: 47°(117°F) Refractive Index: 1.5500 Odor: Pungent Melting Point: -50° Boiling Point: 159° Beilstein Reference: 104505 UN No.: 3073 MDL No.: MFCD00006355

Formula: C₇H₇N
MW: 105.14 g/mol
Boiling Pt: 159 °C (1013 hPa)
Melting Pt: -50 °C
Density: 0.9735 g/cm³ (20 °C)
Flash Pt: 51 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006355
CAS Number: 100-69-6
EINECS: 202-879-8
UN: 3073
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR