2-Methoxybenzyl alcohol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(Hydroxymethyl)anisole, (2-Methoxyphenyl)methanol

A14016-14 A14016-22
AAA14016-14EA 72.3 USD
AAA14016-14 AAA14016-22
2-Methoxybenzyl alcohol 99%
2-Methoxybenzyl alcohol

Purity: 99% CAS No.: 612-16-8 Molecular Formula: C8H10O2 Formula Weight: 138.17 Flash Point: 110°(230°F) Refractive Index: 1.5475 Boiling Point: 248-250° Beilstein Reference: 2043673 MDL No.: MFCD00004611

Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 248…250 °C
Density: 1.12 g/cm³ (20 °C)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004611
CAS Number: 612-16-8
EINECS: 210-296-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR