m-Anisaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Methoxybenzaldehyde
A13962-36
A13962-22
A13962-14
AAAA13962-36EA
416
USD
AAAA13962-36
AAA13962-22
AAA13962-14
m-Anisaldehyde 98%
m-Anisaldehyde
Purity: 98%
CAS No.: 591-31-1
Molecular Formula: C8H8O2
Formula Weight: 136.15
Flash Point: 121°(250°F)
Refractive Index: 1.5530
Odor: Characteristic
Boiling Point: 231-233°
Beilstein Reference: 606013
MDL No.: MFCD00003361
Formula:
C₈H₈O₂ MW: 136.15 g/mol Boiling Pt: 231…233 °C Density: 1.1149 g/cm³ (20 °C) Flash Pt: 121 °C(250 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003361 CAS Number: 591-31-1 EINECS: 209-712-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...