4-Biphenylcarbonyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-Phenylbenzoyl chloride, 4-Biphenylcarbonyl chloride
A13956-18
A13956-09
AAA13956-18EA
198
USD
AAA13956-18
AAA13956-09
4-Biphenylcarbonyl chloride 98%
4-Biphenylcarbonyl chloride
Purity: 98%
CAS No.: 14002-51-8
Molecular Formula: C13H9ClO
Formula Weight: 216.67
Melting Point: 110-115°
Boiling Point: 160°/2mm
UN No.: 3261
MDL No.: MFCD00000692
Formula:
C₁₃H₉ClO MW: 216.67 g/mol Boiling Pt: 160 °C (2 mmHg) Melting Pt: 110…115 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000692 CAS Number: 14002-51-8 EINECS: 237-804-8 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...