Cyclopropanemethylamine 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13946-14
A13946-06
A13946-03
AAA13946-14EA
545.22
USD
AAA13946-14
AAA13946-06
AAA13946-03
Cyclopropanemethylamine 98+%
Cyclopropanemethylamine
Purity: 98+%
CAS No.: 2516-47-4
Molecular Formula: C4H9N
Formula Weight: 71.12
Flash Point: 2°(36°F)
Refractive Index: 1.4330
Odor: Amine-like
Boiling Point: 85-86°
Beilstein Reference: 773674
UN No.: 2733
MDL No.: MFCD00037147
Formula:
C₄H₉N MW: 71.12 g/mol Boiling Pt: 85…86 °C Density: 0.82 g/cm³ (20 °C) Flash Pt: 2 °C (36 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00037147 CAS Number: 2516-47-4 EINECS: 219-737-6 UN: 2733 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...