3-Chlorobenzoylacetonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3'-Chloro-2-cyanoacetophenone, (3-Chlorobenzoyl)acetonitrile, 3-(3-Chlorophenyl)-3-oxopropanenitrile
A13913-03
A13913-06
AAA13913-03EA
27
USD
AAA13913-03
AAAA13913-14
AAA13913-06
3-Chlorobenzoylacetonitrile 98%
3-Chlorobenzoylacetonitrile
Purity: 98%
CAS No.: 21667-62-9
Molecular Formula: C9H6ClNO
Formula Weight: 179.61
Melting Point: 78-83°
Beilstein Reference: 2414476
Formula:
C₉H₆ClNO MW: 179.61 g/mol Melting Pt: 78…83 °C Storage Temperature: Ambient |
MDL Number:
MFCD00067891 CAS Number: 21667-62-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...