2-Butyne 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Purity: 98%
CAS No.: 503-17-3
Molecular Formula: CH3C≡CCH3
Formula Weight: 54.09
Storage Temperature: 2°C to 8°C
Flash Point: -49°(-56°F)
Refractive Index: 1.3920
Odor: Recognizable
Melting Point: -32°
Boiling Point: 24-27°
Beilstein Reference: 1361340
UN No.: 3295
MDL No.: MFCD00009275
Formula:
C₄H₆ Boiling Pt: 24…27 °C Melting Pt: –32 °C Density: 0.690 Flash Pt: –49 °C (–56 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00009275 CAS Number: 503-17-3 EINECS: 207-962-2 UN: 1144 ADR: 3,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...