4-Nitro-3-(trifluoromethyl)aniline 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Amino-2-nitro-α,α,α-trifluorotoluene, 4-Nitro-α,α,α-trifluoro-m-toluidine, 5-Amino-2-nitrobenzotrifluoride

A13798-03 A13798-06
AAA13798-03EA 35.5 USD
AAA13798-03 AAA13798-06
4-Nitro-3-(trifluoromethyl)aniline 98%
4-Nitro-3-(trifluoromethyl)aniline

Purity: 98% CAS No.: 393-11-3 Molecular Formula: C7H5F3N2O2 Formula Weight: 206.13 Melting Point: 125-128° Beilstein Reference: 2650702 UN No.: 2811 MDL No.: MFCD00014717

Formula: C₇H₅F₃N₂O₂
MW: 206.12 g/mol
Melting Pt: 125…128 °C
Density: 1.752 g/cm³ (-123.15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00014717
CAS Number: 393-11-3
EINECS: 206-884-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR