2-Bromopyrimidine 98+%

Supplier: Thermo Scientific Chemicals
Warning

A13791-06 A13791-22 A13791-14
AAA13791-06EA 86.8 USD
AAA13791-06 AAA13791-22 AAA13791-14
2-Bromopyrimidine 98+%
2-Bromopyrimidine

Purity: 98+% CAS No.: 4595-60-2 Molecular Formula: C4H3BrN2 Formula Weight: 158.99 Storage Temperature: 2°C to 8°C Melting Point: 53-57° Boiling Point: 62-64°/1.5mm Beilstein Reference: 605945 MDL No.: MFCD00014601

Formula: C₄H₃BrN₂
MW: 158.99 g/mol
Boiling Pt: 62…64 °C/1.5 mm
Melting Pt: 53…57 °C
Density: Min. 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00014601
CAS Number: 4595-60-2
EINECS: 224-993-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR