Trimethyl orthoformate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13760-AP
A13760-0F
A13760-AE
AAA13760-APEA
66.9
USD
AAA13760-AP
AAA13760-0F
AAAA13760-AE
Trimethyl orthoformate 99%
Trimethyl orthoformate
Purity: 99%
CAS No.: 149-73-5
Molecular Formula: HC(OCH3)3
Formula Weight: 106.12
Flash Point: 15°(59°F)
Refractive Index: 1.3790
Melting Point: -53°
Boiling Point: 101-102°
Beilstein Reference: 969215
Merck Reference: 14,6884
UN No.: 3272
MDL No.: MFCD00008483
Formula:
C₄H₁₀O₃ MW: 106.12 g/mol Boiling Pt: 101 °C (1013 hPa) Melting Pt: -53 °C Density: 0.9691 g/cm³ (20 °C) Flash Pt: 13 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008483 CAS Number: 149-73-5 EINECS: 205-745-7 UN: 3271 ADR: 3,II Merck Index: 12,07012 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...