2,4-Dihydroxybenzophenone 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2,4-Dihydroxyphenyl)(phenyl)methanone, DHBP

A13749-0E A13749-22 A13749-36
AAA13749-0EEA 529 USD
AAA13749-0E AAA13749-22 AAA13749-36
2,4-Dihydroxybenzophenone 99%
2,4-Dihydroxybenzophenone

Purity: 99% CAS No.: 131-56-6 Molecular Formula: C13H10O3 Formula Weight: 214.22 Flash Point: 125°(257°F) Melting Point: 144-147° Boiling Point: 194°/1mm Beilstein Reference: 1311566 Merck Reference: 14,1106 MDL No.: MFCD00002277

Formula: C₁₃H₁₀O₃
MW: 214.22 g/mol
Boiling Pt: 194 °C/1 mm
Melting Pt: 144…147 °C
Density: 0.696 g/cm³ (20 °C)
Flash Pt: 125 °C (257 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002277
CAS Number: 131-56-6
EINECS: 205-029-4
UN: 3077
ADR: 9,III
Merck Index: 13,01107

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR