3-Oxoglutaric acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Acetone-1,3-dicarboxylic acid, 3-Ketoglutaric acid, 3-Oxopentanedioic acid, 1,3-Acetonedicarboxylic acid, β-Ketoglutaric acid

A13742-0B A13742-18 A13742-30
AAA13742-0BEA 554 USD
AAA13742-0B AAA13742-18 AAA13742-30
3-Oxoglutaric acid 97%
3-Oxoglutaric acid

Purity: 97% CAS No.: 542-05-2 Molecular Formula: C5H6O5 Formula Weight: 146.10 Storage Temperature: 2°C to 8°C Melting Point: ca 132° dec. Beilstein Reference: 1447081 Merck Reference: 14,68 MDL No.: MFCD00002711

Formula: C₅H₆O₅
MW: 146.1 g/mol
Boiling Pt: 323 °C (1013 hPa)
Melting Pt: ∼ 132 °C (dec.)
Density: 1.55 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002711
CAS Number: 542-05-2
EINECS: 208-797-9
Merck Index: 13,00069

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR