L(+)-Isoleucine 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
L-Isoleucine, (2S,3S)-(+)-Isoleucine, H-Ile-OH
A13699-09
A13699-18
A13699-22
A13699-30
AAA13699-09EA
58
USD
AAA13699-09
AAA13699-18
AAAA13699-22
AAA13699-30
L(+)-Isoleucine 99%
L(+)-Isoleucine
Purity: 99%
CAS No.: 73-32-5
Molecular Formula: C6H13NO2
Formula Weight: 131.18
Melting Point: ca 287° dec.
Beilstein Reference: 1721792
Merck Reference: 14,5179
Optical Rotation: +40° (c=5 in 5N HCl)
MDL No.: MFCD00064222
Formula:
H₃CCH₂CH(CH₃)CH(NH₂)COOH MW: 131.17 g/mol Melting Pt: ∼ 287 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00064222 CAS Number: 73-32-5 EINECS: 200-798-2 Merck Index: 14,05179 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...