3-Bromoaniline 98%

Supplier: Thermo Scientific Chemicals
Warning

A13665-22 A13665-36 A13665-14
AAA13665-22EA 174 USD
AAA13665-22 AAA13665-36 AAA13665-14
3-Bromoaniline 98%
3-Bromoaniline

Purity: 98% CAS No.: 591-19-5 Molecular Formula: C6H6BrN Formula Weight: 172.03 Flash Point: 110°(230°F) Refractive Index: 1.6250 Odor: Characteristic Melting Point: 16° Boiling Point: 251° Beilstein Reference: 742028 UN No.: 2810 MDL No.: MFCD00007757

Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 251 °C (1013 hPa)
Melting Pt: 16 °C
Density: 1.59 g/cm³ (20 °C)
Flash Pt: ≥ 230 °C
Storage Temperature: Ambient
MDL Number: MFCD00007757
CAS Number: 591-19-5
EINECS: 209-704-4
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR