2-Bromobenzenesulphonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Bromobenzene-1-sulphonyl chloride

A13645-06 A13645-14
AAA13645-06EA 55.2 USD
AAA13645-06 AAA13645-14 AAA13645-03
2-Bromobenzenesulphonyl chloride 98%
2-Bromobenzenesulphonyl chloride

Purity: 98% CAS No.: 2905-25-1 Molecular Formula: C6H4BrClO2S Formula Weight: 255.52 Melting Point: 48-52° Boiling Point: 127-128°/2.4mm UN No.: 3261 MDL No.: MFCD00051971

Formula: C₆H₄BrClO₂S
MW: 255.52
Boiling Pt: 127…128 °C (2.4 mmHg)
Melting Pt: 51
Storage Temperature: Ambient
MDL Number: MFCD00051971
CAS Number: 2905-25-1
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR