o-Toluidine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Toluidine, 2-Aminotoluene, 2-Methylaniline
A13602-0B
A13602-0I
A13602-30
AAA13602-0BEA
60.8
USD
AAA13602-0B
AAA13602-0I
AAA13602-30
o-Toluidine 99%
o-Toluidine
Purity: 99%
CAS No.: 95-53-4
Molecular Formula: C7H9N
Formula Weight: 107.16
Flash Point: 85°(185°F)
Refractive Index: 1.5720
Odor: Unpleasant
Melting Point: -24°
Boiling Point: 199-200°
Beilstein Reference: 741981
Merck Reference: 14,9536
UN No.: 1708
MDL No.: MFCD00007730
Formula:
C₇H₉N MW: 107.16 g/mol Boiling Pt: 200 Melting Pt: –24 °C Density: 1.004 Flash Pt: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007730 CAS Number: 95-53-4 EINECS: 202-429-0 UN: 1708 ADR: 6.1 II Merck Index: 14,9536 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...