3-Fluorophenol 98%

Supplier: Thermo Scientific Chemicals
Danger

A13598-09 A13598-18
AAA13598-09EA 63.12 USD
AAA13598-09 AAA13598-18
3-Fluorophenol 98%
3-Fluorophenol

Purity: 98% CAS No.: 372-20-3 Molecular Formula: C6H5FO Formula Weight: 112.10 Flash Point: 71°(159°F) Refractive Index: 1.5140 Odor: Phenol-like Melting Point: 12-14° Boiling Point: 177-179° Beilstein Reference: 1904536 UN No.: 2922 MDL No.: MFCD00002254

Formula: C₆H₅FO
MW: 112.1 g/mol
Boiling Pt: 178 °C (1013 hPa)
Melting Pt: 13.7 °C
Density: 1.24 g/cm³ (20 °C)
Flash Pt: 71 °C (159 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002254
CAS Number: 372-20-3
EINECS: 206-748-6
UN: 2810
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR