myo-Inositol 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
i-Inositol, D-Myo-Inositol, rel-(1R,2r,3S,4R,5s,6S)-Cyclohexane-1,2,3,4,5,6-hexaol, Inositol
A13586-36
A13586-0E
A13586-22
AAA13586-36EA
204.99
USD
AAA13586-36
AAA13586-0E
AAA13586-22
myo-Inositol 98+%
myo-Inositol
A lipotropic agent
Purity: 98+%
CAS No.: 87-89-8
Molecular Formula: C6H12O6
Formula Weight: 180.16
Physical Form: Cryst powder
Density: 1.75
Appearance: White
Odor: Odorless
Melting Point: 224-227°
Beilstein Reference: 1907329
Merck Reference: 14,4978
MDL No.: MFCD00077932
A lipotropic agent
Formula:
C₆H₁₂O₆ MW: 180.16 g/mol Boiling Pt: 291 °C (1013 hPa) Melting Pt: 210 °C Density: 1.752 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00077932 CAS Number: 87-89-8 EINECS: 201-781-2 Merck Index: 13,05001 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...