9-(Chloromethyl)anthracene 98+%

Supplier: Thermo Scientific Chemicals
Danger

A13579-06 A13579-14
AAA13579-06EA 145 USD
AAA13579-06 AAA13579-14
9-(Chloromethyl)anthracene 98+%
9-(Chloromethyl)anthracene

Purity: 98+% CAS No.: 24463-19-2 Molecular Formula: C15H11Cl Formula Weight: 226.71 Melting Point: 138-142° Beilstein Reference: 1873394 UN No.: 3261 MDL No.: MFCD00001263

Formula: C₁₅H₁₁Cl
MW: 226.71 g/mol
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 138…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00001263
CAS Number: 24463-19-2
EINECS: 246-274-7
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR