2,4-Dihydroxypyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

A13566-06 A13566-14 A13566-03
AAA13566-06EA 191 USD
AAA13566-06 AAAA13566-14 AAA13566-03
2,4-Dihydroxypyridine 97%
2,4-Dihydroxypyridine

Purity: 97% CAS No.: 626-03-9 Molecular Formula: C5H5NO2 Formula Weight: 111.10 Melting Point: ca 267° dec. Beilstein Reference: 108533

Formula: C₅H₅NO₂
Boiling Pt: 315 °C (1013 hPa)
Melting Pt: 275 - 277 °C
Storage Temperature: Ambient
MDL Number: MFCD00006273
CAS Number: 626-03-9
EINECS: 210-924-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR