2-Bromo-1-indanol 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Bromo-1-hydroxyindane

A13511-09 A13511-18
AAA13511-09EA 37.6 USD
AAA13511-09 AAA13511-18 AAA13511-30
2-Bromo-1-indanol 99%
2-Bromo-1-indanol

Purity: 99% CAS No.: 5400-80-6 Molecular Formula: C9H9BrO Formula Weight: 213.08 Melting Point: 128-132° Beilstein Reference: 2046877

Formula: C₉H₉BrO
MW: 213.07 g/mol
Boiling Pt: 290 °C (1013 hPa)
Melting Pt: 128…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00003798
CAS Number: 5400-80-6
EINECS: 226-442-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR