Succinimide 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pyrrolidine-2,5-dione

A13503-0I A13503-0B A13503-30
AAA13503-0IEA 647 USD
AAA13503-0I AAA13503-0B AAA13503-30
Succinimide 98+%
Succinimide

Purity: 98+% CAS No.: 123-56-8 Molecular Formula: C4H5NO2 Formula Weight: 99.09 Flash Point: 201°(393°F) Odor: Odorless pH: 4-6 Melting Point: 121-126° Boiling Point: 287-288° Beilstein Reference: 108440 Merck Reference: 14,8871 MDL No.: MFCD00005495

Formula: C₄H₅NO₂
MW: 99.09 g/mol
Boiling Pt: 287…288 °C
Melting Pt: 121…126 °C
Density: 1.418 g/cm³ (20 °C)
Flash Pt: 201 °C(393 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005495
CAS Number: 123-56-8
EINECS: 204-635-6
Merck Index: 12,09040

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR