Tetrachloro-p-benzoquinone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tetrachloroquinone, 2,3,5,6-Tetrachloro-1,4-benzoquinone, 4-Chloranil, p-Chloranil, 2,3,5,6-Tetrachlorobenzo-1,4-quinone, Tetrachloro-1,4-benzoquinone
A13495-36
A13495-14
A13495-22
AAAA13495-36EA
458
USD
AAAA13495-36
AAA13495-14
AAA13495-22
Tetrachloro-p-benzoquinone 97%
Tetrachloro-p-benzoquinone
Purity: 97%
CAS No.: 118-75-2
Molecular Formula: C6Cl4O2
Formula Weight: 245.88
Melting Point: ca 290° subl.
Beilstein Reference: 393006
Merck Reference: 14,2078
UN No.: 3077
MDL No.: MFCD00001594
Formula:
C₆Cl₄O₂ MW: 245.88 g/mol Boiling Pt: 270 °C (1013 hPa) Melting Pt: ∼ 290 °C (subl.) Density: 1.97 g/cm³ (20 °C) Flash Pt: ≥ 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001594 CAS Number: 118-75-2 EINECS: 204-274-4 UN: 2811 ADR: 6.1,III Merck Index: 13,02088 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...