1,2-Dichloro-4-iodobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,4-Dichloroiodobenzene

A13440-09 A13440-18
AAA13440-30EA 369.74 USD
AAA13440-30 AAA13440-09 AAA13440-18
1,2-Dichloro-4-iodobenzene 98%
1,2-Dichloro-4-iodobenzene

Purity: 98% CAS No.: 20555-91-3 Molecular Formula: C6H3Cl2I Formula Weight: 272.90 Refractive Index: 1.6480 Melting Point: 29-31° Boiling Point: 116-118°/12mm Beilstein Reference: 2245136 MDL No.: MFCD00019014

Formula: C₆H₃Cl₂I
MW: 272.9 g/mol
Boiling Pt: 116…118 °C/12 mm
Melting Pt: 29…31 °C
Storage Temperature: Ambient
MDL Number: MFCD00019014
CAS Number: 20555-91-3
EINECS: 243-874-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR