Cycloheptanone 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Suberon, Cycloheptan-1-one
A13421-18
A13421-30
AAA13421-18EA
48.01
USD
AAA13421-18
AAA13421-30
Cycloheptanone 98+%
Cycloheptanone
Purity: 99%
CAS No.: 502-42-1
Molecular Formula: C7H12O
Formula Weight: 112.17
Flash Point: 55°(131°F)
Refractive Index: 1.4610
Odor: Sweet
Melting Point: -21°
Boiling Point: 179-181°
Beilstein Reference: 969823
Merck Reference: 14,2722
UN No.: 1224
MDL No.: MFCD00004159
Formula:
C₇H₁₂O MW: 112.17 g/mol Boiling Pt: 179…181 °C Melting Pt: –21 °C Density: 0.949 g/cm³ (20 °C) Flash Pt: 55 °C (131 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004159 CAS Number: 502-42-1 EINECS: 207-937-6 UN: 1224 ADR: 3,III Merck Index: 13,02751 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...