Quinaldine 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methylquinoline

A13412-22 A13412-36 A13412-14
AAA13412-22EA 73.7 USD
AAA13412-22 AAA13412-36 AAA13412-14
Quinaldine 96%
Quinaldine

Used in preparation of oil-soluble dyes


It is an anaesthetic used in fish transportation. Also used in anti-malaria drugs, in manufacturing dyes, food colorants , pharmaceuticals and pH indicators.


Precaución: Air sensitive. Light sensitive. Store under argon.

Formula: C₁₀H₉N
MW: 143.19 g/mol
Boiling Pt: 247.6 °C (1013 hPa)
Melting Pt: -2 °C
Density: 1.0585 g/cm³ (20 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00006756
CAS Number: 91-63-4
EINECS: 202-085-1
Merck Index: 12,08228

Order Now

Specification Test Results

Appearance (Color) Clear colorless to yellow to brown
Form Liquid
Assay (GC) ≥95.0%
Refractive Index 1.6080 - 1.6145 @ 20°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR