Phenethylamine 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: PEA, β-Phenylethylamine, 2-Phenylethylamine

A13402-AU A13402-AK A13402-AD
AAA13402-AUEA 206.99 USD
AAA13402-AU AAA13402-AK AAA13402-AD
Phenethylamine 99%
Phenethylamine

Purity: 99% CAS No.: 64-04-0 Molecular Formula: C8H11N Formula Weight: 121.18 Flash Point: 81°(178°F) Refractive Index: 1.5335 Odor: Amine-like pH: 11.5 Melting Point: -60° Boiling Point: 200-202° Beilstein Reference: 507488 Merck Reference: 14,7225 UN No.: 2922 MDL No.: MFCD00008184

Formula: C₆H₅CH₂CH₂NH₂
MW: 121.18 g/mol
Boiling Pt: 200…202 °C
Melting Pt: –60 °C
Density: 0.9639 g/cm³ (20 °C)
Flash Pt: 81 °C (178 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008184
CAS Number: 64-04-0
EINECS: 200-574-4
UN: 2922
ADR: 8,III
Merck Index: 12,07371

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR