Dichlone 98%

Supplier: Thermo Scientific Chemicals
Warning

A13378-22 A13378-36 A13378-14
AAA13378-22EA 56.4 USD
AAA13378-22 AAA13378-36 AAAA13378-14
Dichlone 98%
Dichlone

Purity: 98% CAS No.: 117-80-6 Molecular Formula: C10H4Cl2O2 Formula Weight: 227.05 Melting Point: 193-196° subl. Boiling Point: 275°/2mm Merck Reference: 14,3045 UN No.: 2811 MDL No.: MFCD00001677

Formula: C₁₀H₄Cl₂O₂
Boiling Pt: 275 °C/2 mm
Melting Pt: 193…196 °C (subl.)
Density: 1.44 g/cm³ (16 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001677
CAS Number: 117-80-6
EINECS: 204-210-5
UN: 2811
ADR: 6.1,III
Merck Index: 13,03071

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR