2-Ethylthiophene 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13372-14
A13372-22
AAA13372-14EA
146
USD
AAA13372-14
AAA13372-22
2-Ethylthiophene 99%
2-Ethylthiophene
Purity: 99%
CAS No.: 872-55-9
Molecular Formula: C6H8S
Formula Weight: 112.19
Flash Point: 21°(69°F)
Refractive Index: 1.5120
Odor: Stench
Boiling Point: 133-134°
Beilstein Reference: 105402
UN No.: 1993
MDL No.: MFCD00005461
Formula:
C₆H₈S Boiling Pt: 133…134 °C Density: 0.991 Flash Pt: 21 °C (69 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005461 CAS Number: 872-55-9 EINECS: 212-830-2 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...