Colchicine, dry 95% (dry weight)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 7α-H-Colchicine, (S)-(-)-Colchicine

A13240-06 A13240-14 A13240-03
AAA13240-06EA 377 USD
AAA13240-06 AAA13240-14 AAA13240-03
Colchicine, dry 95% (dry weight)
Colchicine

Purity: 95% CAS No.: 64-86-8 Molecular Formula: C22H25NO6 Formula Weight: 399.44 Melting Point: ca 150° dec. Beilstein Reference: 2228813 Merck Reference: 14,2471 UN No.: 1544 MDL No.: MFCD00078484

Formula: C₂₂H₂₅NO₆
MW: 399.44 g/mol
Melting Pt: ∼ 150 °C (dec.)
Density: 1.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00078484
CAS Number: 64-86-8
EINECS: 200-598-5
UN: 1544
ADR: 6.1,III
Merck Index: 12,02536

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR