4-Methoxyphenylacetone 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-(p-Methoxyphenyl)-2-propanone, 1-(4-Methoxyphenyl)-2-propanone

A13209-22 A13209-14 A13209-36
AAA13209-22EA 138 USD
AAA13209-22 AAA13209-14 AAA13209-36
4-Methoxyphenylacetone 98%
4-Methoxyphenylacetone

Purity: 98% CAS No.: 122-84-9 Molecular Formula: C10H12O2 Formula Weight: 164.20 Flash Point: 101°(213°F) Refractive Index: 1.5250 Boiling Point: 266-268° Beilstein Reference: 2044332 MDL No.: MFCD00008773

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 266…268 °C
Melting Pt: -15 °C
Density: 1.07 g/cm³ (20 °C)
Flash Pt: 101 °C(213 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008773
CAS Number: 122-84-9
EINECS: 204-578-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR