Methyl-2-iodobenzoate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13204-18
A13204-09
AAA13204-18EA
119
USD
AAA13204-18
AAA13204-09
Methyl-2-iodobenzoate 98%
Methyl-2-iodobenzoate
Purity: 98%
CAS No.: 610-97-9
Molecular Formula: C8H7IO2
Formula Weight: 262.05
Flash Point:
Formula:
C₈H₇IO₂ Boiling Pt: 272 °C (1013 hPa) Density: 1.784 g/cm³ (20 °C) Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00016351 CAS Number: 610-97-9 EINECS: 210-243-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...