n-Octane 98+%

Supplier: Thermo Scientific Chemicals
Danger

A13181-AP A13181-0F A13181-AE
AAA13181-APEA 154.15 USD
AAA13181-AP AAA13181-0F AAAA13181-AE
n-Octane 98+%
n-Octane

MDL: MFCD00009556 Beilstein: 1696875 Soluble in ether, Slightly soluble in alcohol


Purity: 98+% CAS No.: 111-65-9 Molecular Formula: CH3(CH2)6CH3 Formula Weight: 114.23 Flash Point: 15°(59°F) Physical Form: Liquid Density: 0.703 Refractive Index: 1.3980 Odor: Nearly odorless Melting Point: -57° Boiling Point: 125-126° Solubility: Soluble in ether, Slightly soluble in alcohol Beilstein Reference: 1696875 Merck Reference: 14,6749 UN No.: 1262 MDL No.: MFCD00009556

Formula: H₃C(CH₂)₆CH₃
MW: 114.23 g/mol
Boiling Pt: 125…126 °C
Melting Pt: –57 °C
Density: 0.703 g/cm³ (20 °C)
Flash Pt: 15 °C (59 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009556
CAS Number: 111-65-9
EINECS: 203-892-1
UN: 1262
ADR: 3,II
Merck Index: 13,06782
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68adf890d68728825d4a2fa4b032700b93f710b8&vid=b00a2925902bc425c80f8ebb3f0e7b456c800ae3&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR