Hexamethyldisilane 98+%

Supplier: Thermo Scientific Chemicals
Danger

A13155-22 A13155-14
AAA13155-22EA 308 USD
AAA13155-22 AAA13155-14
Hexamethyldisilane 98+%
Hexamethyldisilane

Purity: 98+% CAS No.: 1450-14-2 Molecular Formula: (CH3)3SiSi(CH3)3 Formula Weight: 146.38 Flash Point: -1°(30°F) Refractive Index: 1.4220 Odor: Odorless Melting Point: 12-13° Boiling Point: 112-113° Beilstein Reference: 1633463 UN No.: 1993 MDL No.: MFCD00008258

Formula: C₆H₁₈Si₂
Boiling Pt: 112…113 °C
Melting Pt: 12…13 °C
Density: 0.721
Flash Pt: –1 °C (30 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008258
CAS Number: 1450-14-2
EINECS: 215-911-0
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR