3-Fluorobenzenesulphonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Fluorobenzene-1-sulphonyl chloride

A13140-03 A13140-06
AAAA13140-14EA 355.2 USD
AAAA13140-14 AAA13140-03 AAA13140-06
3-Fluorobenzenesulphonyl chloride 98%
3-Fluorobenzenesulphonyl chloride

Purity: 98% CAS No.: 701-27-9 Molecular Formula: C6H4ClFO2S Formula Weight: 194.61 Flash Point: 110°(230°F) Refractive Index: 1.5290 Melting Point: 7° Boiling Point: 231-232° Beilstein Reference: 2091631 UN No.: 3265 MDL No.: MFCD00042286

Formula: C₆H₄ClFO₂S
Boiling Pt: 231…232 °C
Melting Pt: 7 °C
Density: 1.463
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042286
CAS Number: 701-27-9
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR