4-Hydroxybenzyl alcohol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13132-30
A13132-18
A13132-22
AAA13132-30EA
373
USD
AAA13132-30
AAA13132-18
AAAA13132-22
4-Hydroxybenzyl alcohol 99%
4-Hydroxybenzyl alcohol
Purity: 99%
CAS No.: 623-05-2
Molecular Formula: C7H8O2
Formula Weight: 124.14
Melting Point: 114-122°
Boiling Point: 251-253°
MDL No.: MFCD00004658
Formula:
C₇H₈O₂ Boiling Pt: 251…253 °C Melting Pt: 114…122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004658 CAS Number: 623-05-2 EINECS: 210-768-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...