Chlorodimethylsilane 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Dimethylchlorosilane

A13113-AD A13113-AK
AAA13113-ADEA 55.62 USD
AAA13113-AD AAA13113-AK
Chlorodimethylsilane 97%
Chlorodimethylsilane

Purity: 97% CAS No.: 1066-35-9 Molecular Formula: C2H7ClSi Formula Weight: 94.62 Storage Temperature: 2°C to 8°C Flash Point: -28°(-18°F) Refractive Index: 1.3830 Odor: Acrid Melting Point: -111° Boiling Point: 34-36° Beilstein Reference: 1633434 UN No.: 2924 MDL No.: MFCD00000495

Formula: C₂H₇ClSi
MW: 94.62 g/mol
Boiling Pt: 34.6 °C (1013 hPa)
Melting Pt: -111 °C
Density: 0.863 g/cm³ (20 °C)
Flash Pt: -28 °C
Storage Temperature: Ambient
MDL Number: MFCD00000495
CAS Number: 1066-35-9
EINECS: 213-912-0
UN: 2924
ADR: 3,I
REACH: 01-2119486782-25

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR