Cyclobutanone 98% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 0.01%
Purity: 98%
CAS No.: 1191-95-3
Molecular Formula: C4H6O
Formula Weight: 70.09
Flash Point: 10°(50°F)
Refractive Index: 1.4210
Odor: Pleasant
Melting Point: -51°
Boiling Point: 97-99°
Beilstein Reference: 1560289
UN No.: 1224
MDL No.: MFCD00001332
Formula:
C₄H₆O Boiling Pt: 97…99 °C Melting Pt: –51 °C Density: 0.93 g/cm³ (20 °C) Flash Pt: 10 °C (50 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001332 CAS Number: 1191-95-3 EINECS: 214-745-6 UN: 1224 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...