2-Chloro-2-methylpropane 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
tert-Butyl chloride
A13004-AK
A13004-AP
A13004-AU
AAA13004-AKEA
46
USD
AAA13004-AK
AAAA13004-AP
AAA13004-AU
2-Chloro-2-methylpropane 98+%
2-Chloro-2-methylpropane
Purity: 98+%
CAS No.: 507-20-0
Molecular Formula: (CH3)3CCl
Formula Weight: 92.57
Flash Point: -27°(-16°F)
Refractive Index: 1.3848
Odor: Mild
Melting Point: -26°
Boiling Point: 50-52°
Beilstein Reference: 1730872
Merck Reference: 14,1562
UN No.: 1127
MDL No.: MFCD00000816
Formula:
C₄H₉Cl MW: 92.57 g/mol Boiling Pt: 50…52 °C Melting Pt: –26 °C Density: 0.85 g/cm³ (20 °C) Flash Pt: –27 °C (–16 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000816 CAS Number: 507-20-0 EINECS: 208-066-4 UN: 1127 ADR: 3,II Merck Index: 13,01561 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...