2-Nitroresorcinol 98%

Supplier: Thermo Scientific Chemicals
Warning

A12970-06 A12970-22 A12970-14
AAA12970-06EA 39.2 USD
AAA12970-06 AAA12970-22 AAA12970-14
2-Nitroresorcinol 98%
2-Nitroresorcinol

Purity: 98% CAS No.: 601-89-8 Molecular Formula: C6H5NO4 Formula Weight: 155.11 Melting Point: 81-85° Beilstein Reference: 2048819 UN No.: 2811 MDL No.: MFCD00007124

Formula: C₆H₅NO₄
Boiling Pt: 232 °C (1013 hPa)
Melting Pt: 81…85 °C
Density: 1.3526 g/cm³ (96.7 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007124
CAS Number: 601-89-8
EINECS: 210-010-9
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR