Heptylbenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-heptylbenzene, 1-Phenylheptane

A12969-36 A12969-22
AAA12969-36EA 895 USD
AAA12969-36 AAA12969-22
Heptylbenzene 98%
Heptylbenzene

Purity: 98% CAS No.: 1078-71-3 Molecular Formula: C13H20 Formula Weight: 176.30 Flash Point: 95°(203°F) Refractive Index: 1.4850 Melting Point: -48° Boiling Point: 230-233° Beilstein Reference: 1905782 UN No.: 3082 MDL No.: MFCD00009545

Formula: C₁₃H₂₀
MW: 176.3 g/mol
Boiling Pt: 230…233 °C
Melting Pt: –48 °C
Density: 0.86 g/cm³ (20 °C)
Flash Pt: 95 °C (203 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009545
CAS Number: 1078-71-3
EINECS: 214-084-3
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR