Ethyl isobutyrate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Methylpropionic acid ethyl ester

A12966-AP A12966-AE A12966-0F
AAA12966-APEA 68.7 USD
AAA12966-AP AAA12966-AE AAA12966-0F
Ethyl isobutyrate 98%
Ethyl isobutyrate

Purity: 98% CAS No.: 97-62-1 Molecular Formula: (CH3)2CHCO2CH2CH3 Formula Weight: 116.16 Flash Point: 13°(55°F) Refractive Index: 1.3870 Odor: Pleasant Melting Point: -88° Boiling Point: 108-110° Beilstein Reference: 773846 Merck Reference: 14,3814 UN No.: 2385 MDL No.: MFCD00009165

Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 112
Melting Pt: -88
Density: 0.869
Flash Pt: 13 °C (55 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009165
CAS Number: 97-62-1
EINECS: 202-595-4
UN: 2385
ADR: 3 II
Merck Index: 14,3814

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR