Ethyl methanesulfonate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12938-18
A12938-06
A12938-30
A12938-09
AAA12938-18EA
220
USD
AAA12938-18
AAAA12938-06
AAAA12938-30
AAA12938-09
Ethyl methanesulfonate 99%
Ethyl methanesulphonate
Purity: 99%
CAS No.: 62-50-0
Molecular Formula: CH3SO3CH2CH3
Formula Weight: 124.16
Flash Point: 100°(212°F)
Physical Form: Liquid
Density: 1.170
Refractive Index: 1.4180
Sensitivity: Moisture Sensitive
Appearance: Colorless
Odor: Fruit-like
Boiling Point: 85-86°/10mm
Beilstein Reference: 773969
Merck Reference: 14,3827
UN No.: 2810
MDL No.: MFCD00007559
Formula:
C₃H₈O₃S MW: 124.16 g/mol Boiling Pt: 213 °C (1013 hPa) Density: 1.2079 g/cm³ (20 °C) Flash Pt: 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007559 CAS Number: 62-50-0 EINECS: 200-536-7 UN: UN2810 ADR: 6.1,II Merck Index: 13,03859 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...