6-Nitroindole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Nitro-1H-indole

A12927-03 A12927-06
AAA12927-03EA 104 USD
AAA12927-03 AAA12927-06 AAA12927-14
6-Nitroindole 97%
6-Nitroindole

Purity: 97% CAS No.: 4769-96-4 Molecular Formula: C8H6N2O2 Formula Weight: 162.15 Melting Point: 141-145° Beilstein Reference: 135977

Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: 141…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00051497
CAS Number: 4769-96-4
EINECS: 225-311-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR