Thioacetamide 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ethanethioamide
A12926-14
A12926-36
A12926-22
AAA12926-14EA
40.05
USD
AAA12926-14
AAA12926-36
AAA12926-22
Thioacetamide 98%
Thioacetamide
MDL: MFCD00008070
Beilstein: 506006
Soluble in water, ethanol
Purity: 98%
CAS No.: 62-55-5
Molecular Formula: C2H5NS
Formula Weight: 75.13
Melting Point: 110-114°
Solubility: Soluble in water, ethanol
Beilstein Reference: 506006
Merck Reference: 14,9319
MDL No.: MFCD00008070
Formula:
CH₃CSNH₂ MW: 75.13 g/mol Melting Pt: 110…114 °C Density: 1.336 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008070 CAS Number: 62-55-5 EINECS: 200-541-4 Merck Index: 13,09391 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...