2,6-Dibromo-4-nitrophenol 98%

Supplier: Thermo Scientific Chemicals

A12916-30 A12916-14 A12916-22
AAAA12916-30EA 253 USD
AAAA12916-30 AAA12916-14 AAA12916-22
2,6-Dibromo-4-nitrophenol 98%
2,6-Dibromo-4-nitrophenol

Purity: 98% CAS No.: 99-28-5 Molecular Formula: C6H3Br2NO3 Formula Weight: 296.90 Melting Point: ca 145° dec. Beilstein Reference: 1245050

Formula: C₆H₃Br₂NO₃
Melting Pt: 148.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00007334
CAS Number: 99-28-5
EINECS: 202-744-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR