4-(Trifluoromethyl)benzenesulfonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-(Trifluoromethyl)benzene-1-sulphonyl chloride

A12907-06 A12907-14
AAA12907-06EA 93.1 USD
AAA12907-06 AAAA12907-14
4-(Trifluoromethyl)benzenesulfonyl chloride 98%
4-(Trifluoromethyl)benzenesulphonyl chloride

Purity: 98% CAS No.: 2991-42-6 Molecular Formula: C7H4ClF3O2S Formula Weight: 244.62 Flash Point: 110°(230°F) Melting Point: 28-34° Boiling Point: 118-120°/18mm Beilstein Reference: 2113604 UN No.: 3261 MDL No.: MFCD00042422

Formula: C₇H₄ClF₃O₂S
MW: 244.62 g/mol
Boiling Pt: 118…120 °C/18 mm
Melting Pt: 28…34 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042422
CAS Number: 2991-42-6
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR